Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tofacitinib metabolite-1 | 1640971-51-2 | 99.6% | C16H20N6O2 | 10MG
SDP

Catalog No. 5000261934
Encompass_Preferred

Tofacitinib metabolite-1 is the primary human metabolite of the JAK inhibitor tofacitinib. This small-molecule reference standard is supplied as a light-yellow solid at high purity for use in pharmacokinetic, metabolism, and LC-MS analytical method development.

  • CAS number: 1640971-51-2
  • Molecular formula: C16H20N6O2
  • Molecular weight: 328.37 g/mol
  • High purity, approximately 99.6%
  • Appearance: light yellow to yellow solid
  • Soluble in DMSO; recommended 100 mg/mL with ultrasonic assistance
  • Suitable for pharmacokinetic, metabolism, and analytical reference studies
  • Storage: protect from light; solids at 4°C; solutions long-term at -80°C

Catalog No. 50-002-61934 Supplier Medchemexpress LLC Supplier No. HY13633610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tofacitinib metabolite-1 is the primary human metabolite of the JAK inhibitor tofacitinib. This small-molecule reference standard is supplied as a light-yellow solid at high purity for use in pharmacokinetic, metabolism, and LC-MS analytical method development.

  • CAS number: 1640971-51-2
  • Molecular formula: C16H20N6O2
  • Molecular weight: 328.37 g/mol
  • High purity, approximately 99.6%
  • Appearance: light yellow to yellow solid
  • Soluble in DMSO; recommended 100 mg/mL with ultrasonic assistance
  • Suitable for pharmacokinetic, metabolism, and analytical reference studies
  • Storage: protect from light; solids at 4°C; solutions long-term at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.