Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tofacitinib metabolite-1 | 1640971-51-2 | MFCD32693921 | 99.6% | 328.37 g/mol | C16H20N6O2 | 5 MG
SDP

Catalog No. 5000222523
Encompass_Preferred

Tofacitinib metabolite-1 is a research-grade metabolite used for pharmacokinetic and metabolism studies of tofacitinib. It is supplied as a high-purity solid suitable for in vitro and in vivo formulation development, with documented solubility and storage conditions for preclinical use.

  • Metabolite of tofacitinib for pharmacokinetic and metabolism studies.
  • High purity (99.6%).
  • Solid, light yellow to yellow appearance.
  • Soluble in DMSO (100 mg/mL); in vivo formulations ≥2.5 mg/mL with appropriate co-solvents.
  • Molecular formula C16H20N6O2; molecular weight 328.37 g/mol.
  • Available in small research quantities (5 mg, 10 mg; larger sizes by quote).
  • Store solid at 4°C protected from light; in solution store at -80°C for long-term stability.

Catalog No. 50-002-22523 Supplier Medchemexpress LLC Supplier No. HY1363365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tofacitinib metabolite-1 is a research-grade metabolite used for pharmacokinetic and metabolism studies of tofacitinib. It is supplied as a high-purity solid suitable for in vitro and in vivo formulation development, with documented solubility and storage conditions for preclinical use.

  • Metabolite of tofacitinib for pharmacokinetic and metabolism studies.
  • High purity (99.6%).
  • Solid, light yellow to yellow appearance.
  • Soluble in DMSO (100 mg/mL); in vivo formulations ≥2.5 mg/mL with appropriate co-solvents.
  • Molecular formula C16H20N6O2; molecular weight 328.37 g/mol.
  • Available in small research quantities (5 mg, 10 mg; larger sizes by quote).
  • Store solid at 4°C protected from light; in solution store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.