Learn More
Cayman Chemical TofacItInIbcItrate 50mg
SDP
Catalog No. 502951811
Encompass_Preferred
A JAX inhibitor (IC50s = 6.1, 12, and 8 nM for JAK1, JAK2, and JAK3, respectively); selective for JAK1-3 over ROCK2 and Lck (IC50s = 3,400 and 3,870 nM, respectively) as well as 28 additional kinases in enzyme assays (IC50s = >10,000 nM); inhibits IL-2-mediated phosphorylation of JAK3 and STAT5 (30 ng/ml); prevents rejection and prolongs survival in murine and cynomolgus monkey models of heterotopic heart and kidney transplantation
Catalog No. 50-295-1811 Supplier Cayman Chemical Supplier No. 1159850 MG
A JAX inhibitor (IC50s = 6.1, 12, and 8 nM for JAK1, JAK2, and JAK3, respectively); selective for JAK1-3 over ROCK2 and Lck (IC50s = 3,400 and 3,870 nM, respectively) as well as 28 additional kinases in enzyme assays (IC50s = >10,000 nM); inhibits IL-2-mediated phosphorylation of JAK3 and STAT5 (30 ng/ml); prevents rejection and prolongs survival in murine and cynomolgus monkey models of heterotopic heart and kidney transplantation