Learn More
Selleck Chemical LLC Tofacitinib (CP-690550) 10mg 477600-75-2 Tasocitinib
SDP
Catalog No. NC1048781
Encompass_Preferred
Tofacitinib (CP-690550,Tasocitinib) is a novel inhibitor of JAK3 with IC50 of 1 nM in cell-free assays, 20- to 100-fold less potent against JAK2 and JAK1. Tofacitinib inhibits the expression of antiapoptotic BCL-A1 and BCL-XL in human plasmacytoid dendritic cells (PDC) and induced PDC apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1048781 Supplier Selleck Chemical LLC Supplier No. S278910MG
Tofacitinib (CP-690550,Tasocitinib) is a novel inhibitor of JAK3 with IC50 of 1 nM in cell-free assays, 20- to 100-fold less potent against JAK2 and JAK1. Tofacitinib inhibits the expression of antiapoptotic BCL-A1 and BCL-XL in human plasmacytoid dendritic cells (PDC) and induced PDC apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.