Learn More
Selleck Chemical LLC Tofogliflozin(CSG 452) 5mg 1201913-82-7
SDP
Supplier:  Selleck Chemical LLC S85585MG
Encompass_Preferred
Tofogliflozin (CSG 452) is a novel sodium-glucose co-transporter 2(SGLT2) inhibitor with IC50 values of 2.9 nM and 8444 nM for hSGLT2 and hSGLT1, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8363