Learn More
Medchemexpress LLC Tofogliflozin (hydrate) | 1201913-82-7 | MFCD27665464 | 98.4% | C22H28O7 | 10MG
SDP
Catalog No. 5000262009
Encompass_Preferred
Tofogliflozin hydrate is the monohydrate form of tofogliflozin, a potent and selective sodium-glucose cotransporter 2 (SGLT2) inhibitor supplied for research use in biochemical assays and preclinical diabetes studies.
  • Potent and selective SGLT2 inhibition (IC50 ≈2.9 nM).
  • Hydrated molecular form for consistent handling and characterization.
  • High purity (~98.4%) suitable for research applications.
  • Available in small-scale packages compatible with preclinical workflows.
  • Soluble in common laboratory solvents such as DMSO and water for assay preparation.
Catalog No. 50-002-62009 Supplier Medchemexpress LLC Supplier No. HY1341310MG
Tofogliflozin hydrate is the monohydrate form of tofogliflozin, a potent and selective sodium-glucose cotransporter 2 (SGLT2) inhibitor supplied for research use in biochemical assays and preclinical diabetes studies.
  • Potent and selective SGLT2 inhibition (IC50 ≈2.9 nM).
  • Hydrated molecular form for consistent handling and characterization.
  • High purity (~98.4%) suitable for research applications.
  • Available in small-scale packages compatible with preclinical workflows.
  • Soluble in common laboratory solvents such as DMSO and water for assay preparation.