Learn More
Grainger Toggle Valve 3-Pos 1/8 NPT
Catalog No. 502750820
Encompass_Preferred
The Grainger Approved 3-Position Toggle Valve is a 1/8" NPT brass valve with a chrome-plated brass handle. It has 3 positions for directing flow: left port to common port, right port to common port, and closed. It has a maximum pressure rating of 600 PSI and temperature rating of 250°F. This valve can be used for air, water, oil, or inert gas systems. It measures 2-1/2" long x 1-5/16" wide x 1-11/16" high.
Catalog No. 50-275-0820 Supplier Grainger Supplier No. 14F299
The Grainger Approved 3-Position Toggle Valve is a 1/8" NPT brass valve with a chrome-plated brass handle. It has 3 positions for directing flow: left port to common port, right port to common port, and closed. It has a maximum pressure rating of 600 PSI and temperature rating of 250°F. This valve can be used for air, water, oil, or inert gas systems. It measures 2-1/2" long x 1-5/16" wide x 1-11/16" high.