Learn More
Grainger Toggle Valve 4 Way Brass
Catalog No. 502750864
Encompass_Preferred
The Pneumadyne Inc. 4-Way Toggle Valve is a pneumatic directional control valve with a toggle handle that allows the user to direct air flow to four ports. It has a 1/4" NPT female threaded port size and operates at a maximum pressure of 150 PSI. The valve body is made of anodized aluminum and it uses O-ring seals. This 4-way toggle valve can be used for a variety of directional air flow control applications.
Catalog No. 50-275-0864 Supplier Grainger Supplier No. 14F281
The Pneumadyne Inc. 4-Way Toggle Valve is a pneumatic directional control valve with a toggle handle that allows the user to direct air flow to four ports. It has a 1/4" NPT female threaded port size and operates at a maximum pressure of 150 PSI. The valve body is made of anodized aluminum and it uses O-ring seals. This 4-way toggle valve can be used for a variety of directional air flow control applications.