Learn More
Grainger Toggle Valve 4Way 1/8 In NPT
Catalog No. 502750852
Encompass_Preferred
The Pneumadyne Inc. 4-Way Toggle Valve is a pneumatic valve with 4 ports that allows flow between 2 ports at a time. It has a chrome-plated brass body with a toggle lever that switches flow between ports 1 & 2 or ports 3 & 4. The valve has 1/4" NPT female thread ports, operates at up to 150 psi, and measures 2-1/4" long x 1-3/8" wide x 1-1/4" high. This 4-way toggle valve can be used for a variety of directional flow control applications in industrial pneumatic systems.
Catalog No. 50-275-0852 Supplier Grainger Supplier No. 14F292
The Pneumadyne Inc. 4-Way Toggle Valve is a pneumatic valve with 4 ports that allows flow between 2 ports at a time. It has a chrome-plated brass body with a toggle lever that switches flow between ports 1 & 2 or ports 3 & 4. The valve has 1/4" NPT female thread ports, operates at up to 150 psi, and measures 2-1/4" long x 1-3/8" wide x 1-1/4" high. This 4-way toggle valve can be used for a variety of directional flow control applications in industrial pneumatic systems.