Learn More
Grainger Toggle Valve 4Way Brass
Catalog No. 502751004
Encompass_Preferred
The Pneumadyne Inc. 3-Way Toggle Valve is a pneumatic valve with 3 ports that allows flow between 2 ports at a time. It has a chrome-plated brass body with a maximum pressure rating of 150 PSI. The valve features a toggle lever that switches flow between Port 1 and Port 2 or Port 2 and Port 3. It has 1/4" NPT female thread ports and measures 2-1/2" long x 1-3/4" wide x 2" high. This 3-way toggle valve can be used for a variety of industrial pneumatic applications to control air flow.
Catalog No. 50-275-1004 Supplier Grainger Supplier No. 14F291
The Pneumadyne Inc. 3-Way Toggle Valve is a pneumatic valve with 3 ports that allows flow between 2 ports at a time. It has a chrome-plated brass body with a maximum pressure rating of 150 PSI. The valve features a toggle lever that switches flow between Port 1 and Port 2 or Port 2 and Port 3. It has 1/4" NPT female thread ports and measures 2-1/2" long x 1-3/4" wide x 2" high. This 3-way toggle valve can be used for a variety of industrial pneumatic applications to control air flow.