Learn More
Grainger Toggle Valve NC 1/8 In FNPT Brass 3 Way
Catalog No. 502750759
Encompass_Preferred
The Grainger 3-Way Toggle Valve is a brass valve with 1/8" female NPT threaded ports. It has a chrome-plated brass handle and is rated for a maximum pressure of 600 PSI at 150 degrees Fahrenheit. This 3-way valve allows flow between 2 ports at a time and has 3 positions to control flow. It can be used to divert flow or mix fluids. The valve body is made of brass for corrosion resistance. This is an industrial-grade valve for general purpose fluid flow control.
Catalog No. 50-275-0759 Supplier Grainger Supplier No. 14F279
The Grainger 3-Way Toggle Valve is a brass valve with 1/8" female NPT threaded ports. It has a chrome-plated brass handle and is rated for a maximum pressure of 600 PSI at 150 degrees Fahrenheit. This 3-way valve allows flow between 2 ports at a time and has 3 positions to control flow. It can be used to divert flow or mix fluids. The valve body is made of brass for corrosion resistance. This is an industrial-grade valve for general purpose fluid flow control.