Learn More
Grainger Toggle Valve NC 1/8 In FNPT Plastic
Catalog No. 502750863
Encompass_Preferred
The 1/8" NPT Plastic Toggle Valve from Grainger is a manually operated on/off valve made of plastic with brass inserts. It has 1/8" female NPT threaded ports and a toggle handle to open and close the valve. The valve body is made of PVC plastic for corrosion resistance. It has a maximum pressure rating of 150 psi @ 73°F and a maximum temperature rating of 140°F. This valve can be used for a variety of general purpose non-potable water and industrial applications. The toggle handle provides a visual indication of valve position.
Catalog No. 50-275-0863 Supplier Grainger Supplier No. 14F289
The 1/8" NPT Plastic Toggle Valve from Grainger is a manually operated on/off valve made of plastic with brass inserts. It has 1/8" female NPT threaded ports and a toggle handle to open and close the valve. The valve body is made of PVC plastic for corrosion resistance. It has a maximum pressure rating of 150 psi @ 73°F and a maximum temperature rating of 140°F. This valve can be used for a variety of general purpose non-potable water and industrial applications. The toggle handle provides a visual indication of valve position.