Learn More
Grainger Toggle Valve NC 1/8 In NPT
Catalog No. 502750736
Encompass_Preferred
The Grainger Approved Toggle Valve is a bronze ball valve with a toggle handle that rotates 90 degrees to open and close the valve. It has female NPT threaded ends and is available in 1/4", 3/8", and 1/2" sizes. The valve is made of bronze for durability and has a maximum pressure rating of 600 PSI at 70°F. This valve can be used for a variety of general purpose fluid and gas applications. The toggle handle provides quick on/off control.
Catalog No. 50-275-0736 Supplier Grainger Supplier No. 14F295
The Grainger Approved Toggle Valve is a bronze ball valve with a toggle handle that rotates 90 degrees to open and close the valve. It has female NPT threaded ends and is available in 1/4", 3/8", and 1/2" sizes. The valve is made of bronze for durability and has a maximum pressure rating of 600 PSI at 70°F. This valve can be used for a variety of general purpose fluid and gas applications. The toggle handle provides quick on/off control.