Learn More
Grainger Toggle Valve NC 1/8 In NPT Brass
Catalog No. 502750735
Encompass_Preferred
The Pneumadyne Inc. 3-Way Toggle Valve is a pneumatic valve with 3 ports and a toggle handle for switching flow between the ports. It has a 1/4" NPT female thread size, operates at up to 150 psi, and switches between normally closed or normally open configurations. The valve body is made of anodized aluminum with a stainless steel stem and Buna-N seals. It can be used to control air flow for pneumatic cylinders, actuators, and other devices. This 3-way valve routes air between ports for extending, retracting, and exhausting pneumatic cylinders and other automation applications.
Catalog No. 50-275-0735 Supplier Grainger Supplier No. 14F294
The Pneumadyne Inc. 3-Way Toggle Valve is a pneumatic valve with 3 ports and a toggle handle for switching flow between the ports. It has a 1/4" NPT female thread size, operates at up to 150 psi, and switches between normally closed or normally open configurations. The valve body is made of anodized aluminum with a stainless steel stem and Buna-N seals. It can be used to control air flow for pneumatic cylinders, actuators, and other devices. This 3-way valve routes air between ports for extending, retracting, and exhausting pneumatic cylinders and other automation applications.