Learn More
Grainger Toggle Valve Stainless Steel NC
Catalog No. 502750660
Encompass_Preferred
The Grainger 14F296 Toggle Valve is a stainless steel toggle valve with 1/4" NPT female inlet and outlet connections. It has a chrome-plated brass handle and stem, and stainless steel spring. This valve can be used for air, water, oil, and inert gas applications. It has a maximum pressure rating of 1000 PSI and temperature range of -65 to 300°F. The valve body measures 1-1/2" in diameter x 2-1/4" in height. This durable valve allows convenient on/off control of flow.
Catalog No. 50-275-0660 Supplier Grainger Supplier No. 14F296
The Grainger 14F296 Toggle Valve is a stainless steel toggle valve with 1/4" NPT female inlet and outlet connections. It has a chrome-plated brass handle and stem, and stainless steel spring. This valve can be used for air, water, oil, and inert gas applications. It has a maximum pressure rating of 1000 PSI and temperature range of -65 to 300°F. The valve body measures 1-1/2" in diameter x 2-1/4" in height. This durable valve allows convenient on/off control of flow.