Learn More
Cayman Chemical Tol-LIke Receptr 2 Monclon 1ea
SDP
Catalog No. 502955376
Encompass_Preferred
Antigen: CHO cells transfected with human TLR2 cDNA • Host: mouse, clone TL2.1 • Cross Reactivity: (+) human and canine TLR2 • Application(s): FC (intracellular and cell surface), ICC, IHC, IP, and WB • TLR2 interacts with TLR1 and coexpression of TLR1 and TLR2 enhances the NF-kB activation in response to a synthetic lipopeptide. Together, they recognize the lipid configuration of the native mycobacterial lipoprotein as well as several triacylated lipopeptides.
Catalog No. 50-295-5376 Supplier Cayman Chemical Supplier No. 135871 EA
Antigen: CHO cells transfected with human TLR2 cDNA • Host: mouse, clone TL2.1 • Cross Reactivity: (+) human and canine TLR2 • Application(s): FC (intracellular and cell surface), ICC, IHC, IP, and WB • TLR2 interacts with TLR1 and coexpression of TLR1 and TLR2 enhances the NF-kB activation in response to a synthetic lipopeptide. Together, they recognize the lipid configuration of the native mycobacterial lipoprotein as well as several triacylated lipopeptides.