Learn More
Cayman Chemical Tol-LIke Receptr 3 Monclon 1ea
SDP
Catalog No. 502955046
Encompass_Preferred
Antigen: human TLR3 amino acids 55-70 • Host: mouse, clone 40C1285.6 • Cross Reactivity: (+) human and canine TLR3 • Application(s): FC (intracellular and cell surface), ICC, IHC, IP, and WB • TLR3 has a restricted expression pattern being expressed in dendritic cells. The expression of TLR3 in a single cell type may indicate a specific role for this molecule in a restricted setting.
Catalog No. 50-295-5046 Supplier Cayman Chemical Supplier No. 135881 EA
Antigen: human TLR3 amino acids 55-70 • Host: mouse, clone 40C1285.6 • Cross Reactivity: (+) human and canine TLR3 • Application(s): FC (intracellular and cell surface), ICC, IHC, IP, and WB • TLR3 has a restricted expression pattern being expressed in dendritic cells. The expression of TLR3 in a single cell type may indicate a specific role for this molecule in a restricted setting.