Learn More
Cayman Chemical Tol-LIke Receptr 4 Monclon 1ea
SDP
Catalog No. 502955667
Encompass_Preferred
Antigen: Ba/F3 cell line expressing human TLR4 cell surface antigen • Isotype: IgG2aK • Host: mouse, clone HTA125 • Cross Reactivity: (+) human and canine TLR4 • Application(s): FC (cell surface and intracellular), ICC, IP, and neutralization • TLR4 is mediator of innate immunity that is essential for microbial recognition and serves as one of the main mediators of LPS signaling.
Catalog No. 50-295-5667 Supplier Cayman Chemical Supplier No. 135891 EA
Antigen: Ba/F3 cell line expressing human TLR4 cell surface antigen • Isotype: IgG2aK • Host: mouse, clone HTA125 • Cross Reactivity: (+) human and canine TLR4 • Application(s): FC (cell surface and intracellular), ICC, IP, and neutralization • TLR4 is mediator of innate immunity that is essential for microbial recognition and serves as one of the main mediators of LPS signaling.