Learn More
Cayman Chemical Toll-Like Receptor 7 Ligand II, Purity: ≥98%, CAS: 226907-52-4, 1 mg
SDP
Catalog No. 502966767
Encompass_Preferred
An orally bioavailable 8-hydroxyadenine analog that induces IFN production by binding to TLR7 on immune cells; induces IFNα in human peripheral blood mononuclear cells (EC50 = 0.14 µM); reduces HCV in Huh7 cells that carry a HCV replicon
Catalog No. 50-296-6767 Supplier Cayman Chemical Supplier No. 172281 MG
An orally bioavailable 8-hydroxyadenine analog that induces IFN production by binding to TLR7 on immune cells; induces IFNα in human peripheral blood mononuclear cells (EC50 = 0.14 µM); reduces HCV in Huh7 cells that carry a HCV replicon