Learn More
Cayman Chemical Tol-LIke Receptr 8 Monclon 1ea
SDP
Catalog No. 502953689
Encompass_Preferred
Antigen: peptide from human TLR8 within the region of amino acids 750-850 • Host: mouse, clone 44C143 • Cross Reactivity: (+) human and mouse TLR8 • Application(s): FC (intracellular and cell surface), IHC (paraffin-embedded sections), and WB • TLRs mediate innate immunity that is essential for microbial recognition. The TLR8 gene contains three exons, two of which have coding function.
Catalog No. 50-295-3689 Supplier Cayman Chemical Supplier No. 135921 EA
Antigen: peptide from human TLR8 within the region of amino acids 750-850 • Host: mouse, clone 44C143 • Cross Reactivity: (+) human and mouse TLR8 • Application(s): FC (intracellular and cell surface), IHC (paraffin-embedded sections), and WB • TLRs mediate innate immunity that is essential for microbial recognition. The TLR8 gene contains three exons, two of which have coding function.