Learn More
Cayman Chemical Tol-LIke Receptr 9 Monclon 1ea
SDP
Catalog No. 502955653
Encompass_Preferred
Antigen: human TLR9 amino acids 268-284 • Host: mouse, clone 26C593.2 • Cross Reactivity: (+) canine, equine, human, mouse, rat, and Rhesus monkey TLR9 • Application(s): FC (intracellular and cell surface), ICC, IHC (frozen and paraffin), and WB • Gene knockout experiments suggest that TLR9 acts as a receptor for unmethylated CpG dinucleotides in bacterial DNA.TLR9 is highly expressed in spleen.
Catalog No. 50-295-5653 Supplier Cayman Chemical Supplier No. 135931 EA
Antigen: human TLR9 amino acids 268-284 • Host: mouse, clone 26C593.2 • Cross Reactivity: (+) canine, equine, human, mouse, rat, and Rhesus monkey TLR9 • Application(s): FC (intracellular and cell surface), ICC, IHC (frozen and paraffin), and WB • Gene knockout experiments suggest that TLR9 acts as a receptor for unmethylated CpG dinucleotides in bacterial DNA.TLR9 is highly expressed in spleen.