Learn More
Abcam 6-p-Toluidino-2-naphthalenesulfonic acid, potassium salt (TNS-K+), 100MG
Catalog No. 50996133
Encompass_Preferred
MW 351.5 Da, Purity >98%. Fluorescent probe for measuring protein conformational changes and quantitative measurement of protein hydrophobicity processes that modify the exposure of the probe to water. Essentially nonfluorescent in water. Fluorescent when bound to membranes (q.y. ~0.27) or hydrophobic domains of proteins (q.y. ~0.72). Fluorescence enhanced by cyclodextrins. Can be used to provide a sensitive method for separating and analyzing cyclodextrins by capillary electrophoresis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-133 Supplier Abcam Supplier No. AB275213100MG
MW 351.5 Da, Purity >98%. Fluorescent probe for measuring protein conformational changes and quantitative measurement of protein hydrophobicity processes that modify the exposure of the probe to water. Essentially nonfluorescent in water. Fluorescent when bound to membranes (q.y. ~0.27) or hydrophobic domains of proteins (q.y. ~0.72). Fluorescence enhanced by cyclodextrins. Can be used to provide a sensitive method for separating and analyzing cyclodextrins by capillary electrophoresis.
The product is subject to the following: Abcam Restricted Use Statement