Learn More
Medchemexpress LLC Tolazamide | 1156-19-0 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0920
Encompass_Preferred
Tolazamide is an orally active sulfonylurea agent that inhibits sulfonylurea receptor 1 (SUR1) linked to the inwardly rectifying potassium channel. It has anti-diabetic properties, lowering blood glucose and decreasing insulin dose while maintaining adequate metabolic control. Tolazamide is able to improve or normalize hyperglycemia and HbA.
  • Inhibits sulfonylurea receptor 1 (SUR1) linked to the inwardly rectifying potassium channel.
  • Possesses anti-diabetic properties.
  • Lowers blood glucose.
  • Decreases insulin dose while maintaining adequate metabolic control.
  • Improves or normalizes hyperglycemia and HbA.
  • Attenuates gluconeogenesis and ketogenesis in preclinical studies.
  • Soluble in DMSO (≥ 39 mg/mL).
Catalog No. 50-004-50815