Learn More
Medchemexpress LLC Tolazamide | 1156-19-0 | 99.9% | 5 MG
SDP
Catalog No. 5000409314
Encompass_Preferred
Tolazamide (U-17835) is an orally active sulfonylurea agent that inhibits sulfonylurea receptor 1 (SUR1) linked to the inwardly rectifying potassium channel (IC50 = 4.2 μM in HEK293 cells transfected with the human receptor). It has anti-diabetic properties, lowering blood glucose and decreasing insulin dose while maintaining adequate metabolic control. Tolazamide can also improve or normalize hyperglycemia and HbA.
  • Orally active sulfonylurea agent
  • Inhibits sulfonylurea receptor 1 (SUR1)
  • Lowers blood glucose
  • Decreases insulin dose
  • Maintains metabolic control
  • Improves or normalizes hyperglycemia and HbA
  • Attenuates gluconeogenesis and ketogenesis
Catalog No. 50-004-09314 Supplier Medchemexpress LLC Supplier No. HYB09205MG
Tolazamide (U-17835) is an orally active sulfonylurea agent that inhibits sulfonylurea receptor 1 (SUR1) linked to the inwardly rectifying potassium channel (IC50 = 4.2 μM in HEK293 cells transfected with the human receptor). It has anti-diabetic properties, lowering blood glucose and decreasing insulin dose while maintaining adequate metabolic control. Tolazamide can also improve or normalize hyperglycemia and HbA.
  • Orally active sulfonylurea agent
  • Inhibits sulfonylurea receptor 1 (SUR1)
  • Lowers blood glucose
  • Decreases insulin dose
  • Maintains metabolic control
  • Improves or normalizes hyperglycemia and HbA
  • Attenuates gluconeogenesis and ketogenesis