Learn More
Medchemexpress LLC Tolazamide | 1156-19-0 | MFCD00083504 | 99.9% | 100 MG
SDP
Catalog No. 5000408710
Encompass_Preferred
Tolazamide (U-17835) is an orally active sulfonylurea agent that inhibits sulfonylurea receptor 1 (SUR1) linked to the inwardly rectifying potassium channel (IC50 = 4.2 μM in HEK293 cells transfected with the human receptor). It possesses anti-diabetic properties, lowering blood glucose in the sulfonylurea class. Tolazamide decreases insulin dose while maintaining adequate metabolic control and can improve or normalize hyperglycemia and HbA.
  • Orally active sulfonylurea agent
  • Inhibits sulfonylurea receptor 1 (SUR1)
  • Lowers blood glucose
  • Decreases insulin dose while maintaining metabolic control
  • Improves or normalizes hyperglycemia and HbA
  • Attenuates gluconeogenesis and ketogenesis in diabetic rats
Catalog No. 50-004-08710 Supplier Medchemexpress LLC Supplier No. HYB0920100MG
Tolazamide (U-17835) is an orally active sulfonylurea agent that inhibits sulfonylurea receptor 1 (SUR1) linked to the inwardly rectifying potassium channel (IC50 = 4.2 μM in HEK293 cells transfected with the human receptor). It possesses anti-diabetic properties, lowering blood glucose in the sulfonylurea class. Tolazamide decreases insulin dose while maintaining adequate metabolic control and can improve or normalize hyperglycemia and HbA.
  • Orally active sulfonylurea agent
  • Inhibits sulfonylurea receptor 1 (SUR1)
  • Lowers blood glucose
  • Decreases insulin dose while maintaining metabolic control
  • Improves or normalizes hyperglycemia and HbA
  • Attenuates gluconeogenesis and ketogenesis in diabetic rats