Learn More
Medchemexpress LLC Tolazoline hydrochloride | 59-97-2 | 99.9% | C10H13ClN2 | 1 ML
SDP
Catalog No. 5000432321
Encompass_Preferred
Tolazoline hydrochloride (Imidaline hydrochloride) is an α-adrenergic receptor antagonist. It inhibits Noradrenaline-induced cell contraction, modulates vascular resistance, increases arterial pressure, and reverses bradycardia and tachypnea. It can be used to study erectile dysfunction, α2-adrenergic receptor agonist-related poisoning, and skin vascular disease research.
  • α-adrenergic receptor antagonist
  • Inhibits Noradrenaline-induced cell contraction
  • Modulates vascular resistance
  • Increases arterial pressure
  • Reverses bradycardia and tachypnea
  • Used to study erectile dysfunction
  • Used to study α2-adrenergic receptor agonist-related poisoning
  • Used to study skin vascular disease research
Catalog No. 50-004-32321 Supplier Medchemexpress LLC Supplier No. HYA0066A10MM/1ML
Tolazoline hydrochloride (Imidaline hydrochloride) is an α-adrenergic receptor antagonist. It inhibits Noradrenaline-induced cell contraction, modulates vascular resistance, increases arterial pressure, and reverses bradycardia and tachypnea. It can be used to study erectile dysfunction, α2-adrenergic receptor agonist-related poisoning, and skin vascular disease research.
  • α-adrenergic receptor antagonist
  • Inhibits Noradrenaline-induced cell contraction
  • Modulates vascular resistance
  • Increases arterial pressure
  • Reverses bradycardia and tachypnea
  • Used to study erectile dysfunction
  • Used to study α2-adrenergic receptor agonist-related poisoning
  • Used to study skin vascular disease research