Learn More
Cayman Chemical TolbutamIde 50mg
SDP
Catalog No. 502981324
Encompass_Preferred
A SUR1/Kir6.2 channel inhibitor (IC50 = 4.9 µM); selective for SUR1/Kir6.2 over SUR2A/Kir6.2 and SUR2B/Kir6.2 channels (IC50s = 85 and 88 µM, respectively); increases glucose-induced insulin secretion and calcium influx in isolated mouse pancreatic islets; reduces blood glucose levels in a mouse model of STZ-induced diabetes at 80 mg/kg
Catalog No. 50-298-1324 Supplier Cayman Chemical Supplier No. 1988850 MG
A SUR1/Kir6.2 channel inhibitor (IC50 = 4.9 µM); selective for SUR1/Kir6.2 over SUR2A/Kir6.2 and SUR2B/Kir6.2 channels (IC50s = 85 and 88 µM, respectively); increases glucose-induced insulin secretion and calcium influx in isolated mouse pancreatic islets; reduces blood glucose levels in a mouse model of STZ-induced diabetes at 80 mg/kg