Learn More
Medchemexpress LLC Tolbutamide | 64-77-7 | 99.96% | 250 MG
SDP
Catalog No. 5000406515
Encompass_Preferred
Tolbutamide is an orally active KATP inhibitor used in diabetes research. It inhibits cell proliferation, stimulates the exocytosis of glucagon, and has been observed to reduce fetal lethality in mice. This compound is suitable for various in vitro and in vivo studies.
  • Orally active KATP inhibitor
  • Inhibits cell proliferation
  • Stimulates exocytosis of glucagon
  • Reduces fetal lethality in mice
  • Used in diabetes research
  • Reduces glioma cell proliferation
  • Activates mitochondrial-like KATP channels
Catalog No. 50-004-06515 Supplier Medchemexpress LLC Supplier No. 250MGHYB0401R
Tolbutamide is an orally active KATP inhibitor used in diabetes research. It inhibits cell proliferation, stimulates the exocytosis of glucagon, and has been observed to reduce fetal lethality in mice. This compound is suitable for various in vitro and in vivo studies.
  • Orally active KATP inhibitor
  • Inhibits cell proliferation
  • Stimulates exocytosis of glucagon
  • Reduces fetal lethality in mice
  • Used in diabetes research
  • Reduces glioma cell proliferation
  • Activates mitochondrial-like KATP channels