Learn More
Cayman Chemical TolbutamIde-d9 10mg
SDP
Catalog No. 502988804
Encompass_Preferred
An internal standard for the quantification of tolbutamide by GC- or LC-MS
Catalog No. 50-298-8804 Supplier Cayman Chemical Supplier No. 3073810 MG
An internal standard for the quantification of tolbutamide by GC- or LC-MS