Learn More
Medchemexpress LLC Tolbutamide-d9 | 1219794-57-6 | MFCD06658779 | 99.0% | 279.40 g·mol⁻¹ | C12H9D9N2O3S | 10 MG
SDP
Catalog No. 5000298018
Encompass_Preferred
Tolbutamide-d9 is the deuterium-labeled analogue of tolbutamide used as an analytical internal standard and tracer for quantitative assays and method development. It is intended for use in LC-MS, GC-MS, NMR, and pharmacokinetic/ADME studies where an isotopically labeled reference improves accuracy and precision.
  • Deuterium-labeled internal standard for quantitative LC-MS and GC-MS
  • High isotopic and chemical purity for accurate quantitation
  • Suitable for pharmacokinetic and ADME studies
  • Provided in small analytical pack sizes for trace-level work
  • Well characterized for analytical method development and validation
Catalog No. 50-002-98018 Supplier Medchemexpress LLC Supplier No. HYB0401S10MG
Tolbutamide-d9 is the deuterium-labeled analogue of tolbutamide used as an analytical internal standard and tracer for quantitative assays and method development. It is intended for use in LC-MS, GC-MS, NMR, and pharmacokinetic/ADME studies where an isotopically labeled reference improves accuracy and precision.
  • Deuterium-labeled internal standard for quantitative LC-MS and GC-MS
  • High isotopic and chemical purity for accurate quantitation
  • Suitable for pharmacokinetic and ADME studies
  • Provided in small analytical pack sizes for trace-level work
  • Well characterized for analytical method development and validation