Learn More
Cayman Chemical Tolcapone-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1246816-93-2, 1 mg
SDP
Catalog No. 502937178
Encompass_Preferred
An internal standard for the quantification of tolcapone by GC- or LC-MS
Catalog No. 50-293-7178 Supplier Cayman Chemical Supplier No. 286971 MG
An internal standard for the quantification of tolcapone by GC- or LC-MS