Learn More
Medchemexpress LLC Tolclofos-methyl | 57018-04-9 | 98.6% | 250 MG
SDP
Catalog No. 5000414441
Encompass_Preferred
Tolclofos-methyl is a broad-spectrum aromatic hydrocarbon fungicide that is utilized as a seed treatment to provide protection against soil-borne and seed-borne fungal pathogens that lead to seed decay and seedling blights.
  • Broad-spectrum aromatic hydrocarbon fungicide
  • Utilized as a seed treatment
  • Provides protection against soil-borne and seed-borne fungal pathogens
  • Prevents seed decay and seedling blights
  • Intended for research use only
Catalog No. 50-004-14441 Supplier Medchemexpress LLC Supplier No. 250MGHYB2053R
Tolclofos-methyl is a broad-spectrum aromatic hydrocarbon fungicide that is utilized as a seed treatment to provide protection against soil-borne and seed-borne fungal pathogens that lead to seed decay and seedling blights.
  • Broad-spectrum aromatic hydrocarbon fungicide
  • Utilized as a seed treatment
  • Provides protection against soil-borne and seed-borne fungal pathogens
  • Prevents seed decay and seedling blights
  • Intended for research use only