Learn More
Medchemexpress LLC Tolebrutinib | 1971920-73-6 | 99.8% | 455.51 | 1 MG
SDP
Catalog No. 5000345054
Encompass_Preferred
Tolebrutinib (SAR442168) is a potent, selective, orally active, and brain-penetrant inhibitor of Bruton tyrosine kinase (BTK), with IC50s of 0.4 and 0.7 nM in Ramos B cells and in HMC microglia cells, respectively. It exhibits efficacy in central nervous system immunity and can be used for the research of multiple sclerosis (MS).
  • Potent, selective, orally active, and brain-penetrant inhibitor of Bruton tyrosine kinase (BTK).
  • Exhibits efficacy in central nervous system immunity.
  • Can be used for the research of multiple sclerosis (MS).
  • High purity: 99.79%.
Catalog No. 50-003-45054 Supplier Medchemexpress LLC Supplier No. HY10919210MM/1ML
Tolebrutinib (SAR442168) is a potent, selective, orally active, and brain-penetrant inhibitor of Bruton tyrosine kinase (BTK), with IC50s of 0.4 and 0.7 nM in Ramos B cells and in HMC microglia cells, respectively. It exhibits efficacy in central nervous system immunity and can be used for the research of multiple sclerosis (MS).
  • Potent, selective, orally active, and brain-penetrant inhibitor of Bruton tyrosine kinase (BTK).
  • Exhibits efficacy in central nervous system immunity.
  • Can be used for the research of multiple sclerosis (MS).
  • High purity: 99.79%.