Learn More
ATCC Toledo; Non-Hodgkin's Lymphoma; Human (Homo sapiens)
Supplier:  ATCC CRL2631
Encompass
Toledo is a B lymphocyte cell line that was isolated in 1990 from the peripheral blood of a White, adult female patient with diffuse large cell lymphoma, non-Hodgkin's B cell. This cell line was deposited by H Ben-Bassat and can be used in immunology research.
Catalog No. 50-238-4279