Learn More
Apexbio Technology LLC Tolfenamic Acid 13710-19-5 10g
SDP
Catalog No. 5000092185
Encompass_Preferred
Tolfenamic acid (CAS 13710-19-5) is a non-steroidal anti-inflammatory drug (NSAID) that functions primarily through the inhibition of cyclooxygenase (COX) enzymes resulting in reduced prostaglandin synthesis and inflammation In cellular studies tolfenamic acid has been observed to inhibit cell proliferation and promote apoptosis as indicated by heightened caspase activity and alterations in bioelectric impedance measurements These findings support its utility in research investigating inflammatory pathways apoptosis induction and the cellular effects of NSAIDs in various biological systems
Catalog No. 50-000-92185 Supplier Apexbio Technology LLC Supplier No. B145510000
Tolfenamic acid (CAS 13710-19-5) is a non-steroidal anti-inflammatory drug (NSAID) that functions primarily through the inhibition of cyclooxygenase (COX) enzymes resulting in reduced prostaglandin synthesis and inflammation In cellular studies tolfenamic acid has been observed to inhibit cell proliferation and promote apoptosis as indicated by heightened caspase activity and alterations in bioelectric impedance measurements These findings support its utility in research investigating inflammatory pathways apoptosis induction and the cellular effects of NSAIDs in various biological systems