Learn More
Medchemexpress LLC Tolfenamic acid | 13710-19-5 | 99.8% | 25 MG
SDP
Catalog No. 5000450528
Encompass_Preferred
Tolfenamic acid is the analytical standard of Tolfenamic acid. It is a non-steroidal anti-inflammatory and anti-cancer agent that selectively inhibits COX-2, with an IC50 of 13.49 μM in LPS-treated canine DH82 monocyte/macrophage cells, but shows no effect on COX-1. This product is intended for research and analytical applications.
  • Analytical standard of Tolfenamic acid
  • Intended for research and analytical applications
  • Non-steroidal anti-inflammatory and anti-cancer agent
  • Selectively inhibits COX-2
Catalog No. 50-004-50528 Supplier Medchemexpress LLC Supplier No. 25MGHYB0335R
Add to Cart
Add to Cart
Tolfenamic acid is the analytical standard of Tolfenamic acid. It is a non-steroidal anti-inflammatory and anti-cancer agent that selectively inhibits COX-2, with an IC50 of 13.49 μM in LPS-treated canine DH82 monocyte/macrophage cells, but shows no effect on COX-1. This product is intended for research and analytical applications.
  • Analytical standard of Tolfenamic acid
  • Intended for research and analytical applications
  • Non-steroidal anti-inflammatory and anti-cancer agent
  • Selectively inhibits COX-2