Learn More
Medchemexpress LLC Tolfenamic acid-13C6 | 1420043-61-3 | 99.7% | 1 MG
SDP
Catalog No. 5000406798
Encompass_Preferred
Tolfenamic acid-13C6 is an isotope-labeled form of Tolfenamic Acid, a non-steroidal anti-inflammatory and anti-cancer agent. This labeled compound is designed for use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS studies.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS
  • Derived from Tolfenamic Acid, a non-steroidal anti-inflammatory and anti-cancer agent
  • Selectively inhibits COX-2
Catalog No. 50-004-06798 Supplier Medchemexpress LLC Supplier No. HYB0335S11MG
Add to Cart
Add to Cart
Tolfenamic acid-13C6 is an isotope-labeled form of Tolfenamic Acid, a non-steroidal anti-inflammatory and anti-cancer agent. This labeled compound is designed for use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS studies.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS
  • Derived from Tolfenamic Acid, a non-steroidal anti-inflammatory and anti-cancer agent
  • Selectively inhibits COX-2