Learn More
Cayman Chemical Tolfenamic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1246820-82-5, 5 mg
SDP
Catalog No. 502937072
Encompass_Preferred
An internal standard for the quantification of tolfenamic acid by GC- or LC-MS
Catalog No. 50-293-7072 Supplier Cayman Chemical Supplier No. 287005 MG
An internal standard for the quantification of tolfenamic acid by GC- or LC-MS