Learn More
Medchemexpress LLC Tolfenamic acid (standard) | 13710-19-5 | 50 MG
SDP
Catalog No. 5000406767
Encompass_Preferred
Tolfenamic Acid (Standard) is primarily used for research and analytical applications. It acts as a non-steroidal anti-inflammatory and anti-cancer agent.
  • Selectively inhibits COX-2, with an IC50 of 13.49 μM in LPS-treated canine DH82 monocyte/macrophage cells
  • Shows no effect on COX-1
  • Intended for research and analytical use
Catalog No. 50-004-06767 Supplier Medchemexpress LLC Supplier No. HYB0335R50MG
Add to Cart
Add to Cart
Tolfenamic Acid (Standard) is primarily used for research and analytical applications. It acts as a non-steroidal anti-inflammatory and anti-cancer agent.
  • Selectively inhibits COX-2, with an IC50 of 13.49 μM in LPS-treated canine DH82 monocyte/macrophage cells
  • Shows no effect on COX-1
  • Intended for research and analytical use