Learn More
TARGETMOL CHEMICALS INC TOLIMIDONE 25MG
SDP
Catalog No. 502576421
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tolimidone (NSC 314335) is a chemical compound which inhibits acid secretion in animal models and also acts as a bronchodilator in histamine-challenged animals. purity: 100%
Catalog No. 50-257-6421 Supplier TARGETMOL CHEMICALS INC Supplier No. T689625MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tolimidone (NSC 314335) is a chemical compound which inhibits acid secretion in animal models and also acts as a bronchodilator in histamine-challenged animals. purity: 100%