Learn More
Medchemexpress LLC Tolinapant | 1799328-86-1 | 99.81% | 539.68 | 1 ML
SDP
Catalog No. 5000345161
Encompass_Preferred
Tolinapant (ASTX660) is an orally bioavailable dual antagonist of cellular inhibitor of apoptosis protein (cIAP) and X-linked inhibitor of apoptosis protein (XIAP).
  • Orally bioavailable dual antagonist of cellular inhibitor of apoptosis protein (cIAP) and X-linked inhibitor of apoptosis protein (XIAP)
  • Being investigated in a single-agent Phase 1/2 clinical trial in patients with advanced solid tumors and lymphomas
  • Demonstrates antiproliferative activity against human MDA-MB-231 cells with an IC50 of 1.8 nM
  • Shows efficacy in HCC1806 xenografts in mice by causing moderate tumor growth inhibition
  • Available with a purity of 99.81%
Catalog No. 50-003-45161 Supplier Medchemexpress LLC Supplier No. HY10956550MG
Tolinapant (ASTX660) is an orally bioavailable dual antagonist of cellular inhibitor of apoptosis protein (cIAP) and X-linked inhibitor of apoptosis protein (XIAP).
  • Orally bioavailable dual antagonist of cellular inhibitor of apoptosis protein (cIAP) and X-linked inhibitor of apoptosis protein (XIAP)
  • Being investigated in a single-agent Phase 1/2 clinical trial in patients with advanced solid tumors and lymphomas
  • Demonstrates antiproliferative activity against human MDA-MB-231 cells with an IC50 of 1.8 nM
  • Shows efficacy in HCC1806 xenografts in mice by causing moderate tumor growth inhibition
  • Available with a purity of 99.81%