Learn More
Cayman Chemical TolLIke Receptr 12 Polyclo 1ea
SDP
Catalog No. 502955666
Encompass_Preferred
Antigen: mouse TLR12 amino acids 911-926 • Host: rabbit • Cross Reactivity: (+) mouse and rat TLR12 • Application(s): FC (intracellular) and WB • TLR12 displays a distinct pattern of expression in macrophages, liver, kidney, and bladder epithelial cells. TLR12 does not respond to known TLR ligands.
Catalog No. 50-295-5666 Supplier Cayman Chemical Supplier No. 135861 EA
Antigen: mouse TLR12 amino acids 911-926 • Host: rabbit • Cross Reactivity: (+) mouse and rat TLR12 • Application(s): FC (intracellular) and WB • TLR12 displays a distinct pattern of expression in macrophages, liver, kidney, and bladder epithelial cells. TLR12 does not respond to known TLR ligands.