Learn More
Cayman Chemical TolLIke RECPTR 7 LIgnd I 500ug
SDP
Catalog No. 502965124
Encompass_Preferred
An orally bioavailable 8-hydroxyadenine analog that induces IFN production by binding to TLR7 on immune cells; induces IFNα in human peripheral blood mononuclear cells (EC50 = 0.14 µM); reduces HCV in Huh7 cells that carry a HCV replicon
Catalog No. 50-296-5124 Supplier Cayman Chemical Supplier No. 17228500 UG
An orally bioavailable 8-hydroxyadenine analog that induces IFN production by binding to TLR7 on immune cells; induces IFNα in human peripheral blood mononuclear cells (EC50 = 0.14 µM); reduces HCV in Huh7 cells that carry a HCV replicon