Learn More
Aviva Systems Biology Corp TOLLIP Antibody - C-terminal region (ARP31758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOLLIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TOLLIP. Alternate names: FLJ33531, IL-1RAcPIPApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54322