Learn More
Medchemexpress LLC HY-B1489 100mg Medchemexpress, Tolmetin (sodium dihydrate) CAS:64490-92-2 Purity:>98%
SDP
Catalog No. 501932952
Encompass_Preferred
Medchemexpress, HY-B1489 100mg Tolmetin (sodium dihydrate) CAS:64490-92-2 Tolmetin sodium dihydrate is a non-steroidal antiinflammatory agent, and acts as a non-selective COX inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2952 Supplier Medchemexpress LLC Supplier No. HYB1489100MG
Medchemexpress, HY-B1489 100mg Tolmetin (sodium dihydrate) CAS:64490-92-2 Tolmetin sodium dihydrate is a non-steroidal antiinflammatory agent, and acts as a non-selective COX inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.