Learn More
Medchemexpress LLC Tolmetin sodium dihydrate | 64490-92-2 | 99.94% | 1 ML
SDP
Catalog No. 5000411376
Encompass_Preferred
Tolmetin sodium dihydrate is an orally active and potent COX inhibitor, with IC50s of 0.35 μM and 0.82 μM for human COX-1 and COX-2, respectively. It is a non-steroidal anti-inflammatory drug (NSAID).
  • Orally active and potent COX inhibitor.
  • Non-steroidal anti-inflammatory drug (NSAID).
  • Shows anticancer activity against HT-29 colon cancer cell line in a dose-dependent manner.
  • Can attenuate quinolinic acid (QA)-induced neurotoxicity.
Catalog No. 50-004-11376 Supplier Medchemexpress LLC Supplier No. HYB148910MM/1ML
Tolmetin sodium dihydrate is an orally active and potent COX inhibitor, with IC50s of 0.35 μM and 0.82 μM for human COX-1 and COX-2, respectively. It is a non-steroidal anti-inflammatory drug (NSAID).
  • Orally active and potent COX inhibitor.
  • Non-steroidal anti-inflammatory drug (NSAID).
  • Shows anticancer activity against HT-29 colon cancer cell line in a dose-dependent manner.
  • Can attenuate quinolinic acid (QA)-induced neurotoxicity.