Learn More
Cayman Chemical TolmetInsodIum salt hydrAT 25g
SDP
Catalog No. 502966550
Encompass_Preferred
An NSAID that non-selectively inhibits human COX-1 and -2 with IC50 values of 0.35 and 0.82 µM, respectively; also inhibits aldo-keto reductases (e.g., IC50 = 2.39 µM for human aldose reductase)
Catalog No. 50-296-6550 Supplier Cayman Chemical Supplier No. 1819525 G
An NSAID that non-selectively inhibits human COX-1 and -2 with IC50 values of 0.35 and 0.82 µM, respectively; also inhibits aldo-keto reductases (e.g., IC50 = 2.39 µM for human aldose reductase)