Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolnaftate, 99.3%, MP Biomedicals™

Catalog No. ICN15693401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15693401 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15693401 Supplier MP Biomedicals, Inc Supplier No. 0215693401

missing translation for 'validMainframeMsg'

Specifications

CAS 2398-96-1
Color White to Yellow
Assay Percent Range 99.3%
Molecular Formula C19H17NOS
Quantity 1 g
Synonym tolnaftate, tolnaphthate, tinactin, tonoftal, sporiline, tinaderm, tolsanil, aftate, chinofungin, dermoxin
InChI Key FUSNMLFNXJSCDI-UHFFFAOYSA-N
SMILES CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2
IUPAC Name O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate
Molecular Weight (g/mol) 307.411
PubChem CID 5510
ChEBI CHEBI:9620
Formula Weight 307.4
Physical Form Crystalline Powder
Chemical Name or Material Tolnaftate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.