Learn More
Cayman Chemical Tolperisone (hydrochloride), Purity: ≥98%, CAS: 3644-61-9, 250 mg
SDP
Catalog No. 502935393
Encompass_Preferred
An inhibitor of voltage-gated sodium channels (IC50 = 198 µM for inhibition of peak sodium currents evoked by step depolarizations in DRG neurons in vitro); acts as an antispastic agent that attenuates monosynaptic, disynaptic, and polysynaptic reflux response to dorsal root stimulation in vivo
Catalog No. 50-293-5393 Supplier Cayman Chemical Supplier No. 26078250 MG
An inhibitor of voltage-gated sodium channels (IC50 = 198 µM for inhibition of peak sodium currents evoked by step depolarizations in DRG neurons in vitro); acts as an antispastic agent that attenuates monosynaptic, disynaptic, and polysynaptic reflux response to dorsal root stimulation in vivo