Learn More
TARGETMOL CHEMICALS INC TOLRESTAT 5MG
SDP
Catalog No. 502581124
Encompass_Preferred
Also available in 2 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tolrestat (AY-27773) is a potent inhibitor of aldose reductase (IC50 = 35 nM). purity: 99%
Catalog No. 50-258-1124 Supplier TARGETMOL CHEMICALS INC Supplier No. T171145MG
Also available in 2 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tolrestat (AY-27773) is a potent inhibitor of aldose reductase (IC50 = 35 nM). purity: 99%